Thermo Scientific™

TMEA (tris(2-maleimidoethyl)amine)

Catalog number: 33043
Have Questions?
Thermo Scientific™

TMEA (tris(2-maleimidoethyl)amine)

Catalog number: 33043
Have Questions?
Catalog Number
33043
Unit Size
50 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
3304350 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce TMEA is a trifunctional crosslinker containing three reactive maleimide groups to enable three-way conjugation and polymerization of sulfhydryl-containing molecules.

Features of Thermo Scientific Pierce TMEA:

• Reactive groups: maleimide (on three termini)
• Reactive towards: sulfhydryl groups
• 3-way, sulfhydryl-to-sulfhydryl crosslinker; tris (2-maleimidoethyl) amine, composed of three maleimide groups on short spacer arms
• Water-insoluble—dissolve first in DMF or DMSO, then add to aqueous reaction buffers
• Noncleavable
• Useful for the preparation of trimeric complexes of cysteine-containing peptides and other sulfhydryl-containing compounds
• Core molecule for the construction of dendritic polymers

Product References:
Crosslinker Application Guide -- search for recent literature references for this product

Related Products
BMH (bismaleimidohexane)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Sulfhydryl-Sulfhydryl
Cleavable
No
Description
TMEA
Molecular Weight (g/mol)
386.36
PEGylated
No
Spacer Arm Length
10.3 Å
Cell Permeability
Yes
Shipping Condition
Ambient
Product Line
Pierce™
Labeling Method
Chemical Labeling
Crosslinker Type
Homotrifunctional
Reactive Moiety
Maleimide
Spacer
Medium (10 to 30 Å)
Form
Solid
Quantity
50 mg
Solubility
DMF, DMSO
Format
Standard
Water Soluble
No
Product Type
Crosslinker

Contents & Storage

Upon receipt store at 4°C protected from moisture.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text