Thermo Scientific Chemicals

Methyl 3-methoxyacrylate, 95%, Thermo Scientific Chemicals

Catalog number: 330461000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 3-methoxyacrylate, 95%, Thermo Scientific Chemicals

Catalog number: 330461000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 330461000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
34846-90-7
IUPAC Name
methyl 3-methoxyprop-2-enoate
Molecular Formula
C5H8O3
InChI Key
AUTCCPQKLPMHDN-UHFFFAOYSA-N
SMILES
COC=CC(=O)OC
Infrared spectrum
Conforms
GC
>=94 %
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Refractive index
1.4490 to 1.4510 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text