Bis(pinacolato)diboron, 98%, Thermo Scientific Chemicals
Bis(pinacolato)diboron, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(pinacolato)diboron, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 330570010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS73183-34-3
IUPAC Name4,4,5,5-tetramethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane
Molecular FormulaC12H24B2O4
InChI KeyIPWKHHSGDUIRAH-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting point135°C to 140°C
Appearance (Form)Powder or platelets
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only