Thermo Scientific™

TSAT (tris-(succinimidyl)aminotriacetate)

Catalog number: 33063
Have Questions?
Thermo Scientific™

TSAT (tris-(succinimidyl)aminotriacetate)

Catalog number: 33063
Have Questions?
Catalog Number
33063
Unit Size
50 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
3306350 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce TSAT is tris-(succinimidyl) aminotriacetate, a novel trifunctional crosslinker that contains three short spacers arms linked to a central nitrogen atom, each ending in an amine-reactive NHS-ester group.

Features of TSAT:

Reactive groups: NHS ester (three ends)
Reactive towards: amino groups (primary amines)
• NHS ester couples to amines at pH 7-9 form stable amide bonds
• Trifunctional amine-reactive crosslinking agent
• Non-cleavable
• Water-insoluble (dissolve first in DMF or DMSO)

Applications:
• Preparation of multicomponent aggregates
• Polyerization of protein interactions for nearest-neighbor studies
• Core molecule for the construction of dendritic polymers

Product References:
Crosslinker Application Guide -- search for recent literature references for this product

Related Products
TSAT (tris-(succinimidyl)aminotriacetate)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Amine-Amine
Cleavable
No
Description
TSAT
Molecular Weight (g/mol)
482.36
PEGylated
No
Spacer Arm Length
4.2 Å
Cell Permeability
Yes
Shipping Condition
Ambient
Product Line
Pierce™
Labeling Method
Chemical Labeling
Crosslinker Type
Homotrifunctional
Reactive Moiety
NHS Ester
Spacer
Short (<10 Å)
Form
Solid
Quantity
50 mg
Solubility
DMF, DMSO
Format
Standard
Water Soluble
No
Product Type
Crosslinker

Contents & Storage

Upon receipt store at 4°C protected from moisture.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text