Thermo Scientific Chemicals

5-Bromo-4-chloro-3-indolyl-beta-D-glucuronide cyclohexylammonium salt hydrate, 98%, Thermo Scientific Chemicals

Catalog number: 331191000
100 mg, Glass bottle, Each
Thermo Scientific Chemicals

5-Bromo-4-chloro-3-indolyl-beta-D-glucuronide cyclohexylammonium salt hydrate, 98%, Thermo Scientific Chemicals

Catalog number: 331191000
100 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 331191000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
114162-64-0
IUPAC Name
(2S,3S,4S,5R,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid; cyclohexanamine
Molecular Formula
C20H26BrClN2O7
InChI Key
JXCKZXHCJOVIAV-VLQIOUPANA-N
SMILES
NC1CCCCC1.O[C@@H]1[C@@H](O)[C@H](OC2=CNC3=CC=C(Br)C(Cl)=C23)O[C@@H]([C@H]1O)C(O)=O
Appearance (Form)
Powder or crystals or needles
HPLC
>=97.5 %
Appearance (Color)
Colorless to white to off-white
Infrared spectrum
Conforms
Water
=<5 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text