2,6-Dimethoxy-1,4-benzoquinone, 97%, Thermo Scientific Chemicals
2,6-Dimethoxy-1,4-benzoquinone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Dimethoxy-1,4-benzoquinone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 331230050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS530-55-2
IUPAC Name2,6-dimethoxycyclohexa-2,5-diene-1,4-dione
Molecular FormulaC8H8O4
InChI KeyOLBNOBQOQZRLMP-UHFFFAOYSA-N
SMILESCOC1=CC(=O)C=C(OC)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Color)Yellow
Appearance (Form)Crystalline powder
Melting point253°C to 257°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only