N-Palmitoyl-D-erythro-sphingosine, synthetical, 98% (99+% e.e.), Thermo Scientific Chemicals
N-Palmitoyl-D-erythro-sphingosine, synthetical, 98% (99+% e.e.), Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Palmitoyl-D-erythro-sphingosine, synthetical, 98% (99+% e.e.), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
25 mg
Packaging:
Glass bottle
Catalog number 331540050
Price (USD)/ Each
-
Quantity:
5 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS24696-26-2
IUPAC NameN-(1,3-dihydroxyoctadec-4-en-2-yl)hexadecanamide
Molecular FormulaC34H67NO3
InChI KeyYDNKGFDKKRUKPY-UHFFFAOYNA-N
SMILESCCCCCCCCCCCCCCCC(=O)NC(CO)C(O)C=CCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Crystalline powder
HPLC>=97.50 %
Enantiomeric excess>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only