Thermo Scientific Chemicals

1-Methylindole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: 331610010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Methylindole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: 331610010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 331610010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
19012-03-4
IUPAC Name
1-methyl-1H-indole-3-carbaldehyde
Molecular Formula
C10H9NO
InChI Key
KXYBYRKRRGSZCX-UHFFFAOYSA-N
SMILES
CN1C=C(C=O)C2=CC=CC=C12
Appearance (Color)
Light yellow to orange to brown
Infrared spectrum
Conforms
GC
>=96.0 %
Appearance (Form)
Crystalline powder and/or chunks
Melting point
67°C to 72°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text