1-Methylindole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals
1-Methylindole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Methylindole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 331610100
Price (USD)/ Each
125.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
148.00 
Save 22.35 (15%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
125.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
148.00 
Save 22.35 (15%)
Add to cart
1-Methylindole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals
Catalog number331610100
Price (USD)/ Each
125.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
148.00 
Save 22.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name1-methyl-1H-indole-3-carbaldehyde
Molecular FormulaC10H9NO
InChI KeyKXYBYRKRRGSZCX-UHFFFAOYSA-N
SMILESCN1C=C(C=O)C2=CC=CC=C12
Molecular Weight (g/mol)159.19
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder and/or chunks
Melting point67°C to 72°C
Appearance (Color)Light yellow to orange to brown
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only