Thermo Scientific Chemicals

(R)-(+)-2-Methoxypropionic acid, 97%, Thermo Scientific Chemicals

Catalog number: 331810010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

(R)-(+)-2-Methoxypropionic acid, 97%, Thermo Scientific Chemicals

Catalog number: 331810010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 331810010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
23943-96-6
IUPAC Name
(2R)-2-methoxypropanoate
Molecular Formula
C4H7O3
InChI Key
ICPWFHKNYYRBSZ-GSVOUGTGSA-M
SMILES
CO[C@H](C)C([O-])=O
Infrared spectrum
Conforms
Refractive index
1.4130 to 1.4150 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow
GC
>=96.0 %
Specific optical rotation
+76° to +81° (20°C, 589 nm) (neat)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text