4-Chloro-o-phenylenediamine, 97%, Thermo Scientific Chemicals
4-Chloro-o-phenylenediamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloro-o-phenylenediamine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 331860250
Price (USD)/ Each
63.65
Online exclusive
70.20 
Save 6.55 (9%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
63.65
Online exclusive
70.20 
Save 6.55 (9%)
Add to cart
4-Chloro-o-phenylenediamine, 97%, Thermo Scientific Chemicals
Catalog number331860250
Price (USD)/ Each
63.65
Online exclusive
70.20 
Save 6.55 (9%)
-
Add to cart
Chemical Identifiers
IUPAC Name4-chlorobenzene-1,2-diamine
Molecular FormulaC6H7ClN2
InChI KeyBXIXXXYDDJVHDL-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C=C1N
Molecular Weight (g/mol)142.59
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Powder or granules and/or chunks
Melting point70°C to 75°C
Appearance (Color)Grey-brown to dark brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only