Thermo Scientific Chemicals

1-Bromo-3,5-dichlorobenzene, 98%, Thermo Scientific Chemicals

Catalog number: 331900250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Bromo-3,5-dichlorobenzene, 98%, Thermo Scientific Chemicals

Catalog number: 331900250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 331900250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
19752-55-7
IUPAC Name
1-bromo-3,5-dichlorobenzene
Molecular Formula
C6H3BrCl2
InChI Key
DZHFFMWJXJBBRG-UHFFFAOYSA-N
SMILES
ClC1=CC(Br)=CC(Cl)=C1
Appearance (Color)
White to beige to light brown
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
74°C to 77°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text