Thermo Scientific Chemicals

Glycerol, 99+%, pure, synthetic, Thermo Scientific Chemicals

Catalog number: 332030025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Glycerol, 99+%, pure, synthetic, Thermo Scientific Chemicals

Catalog number: 332030025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 332030025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-81-5
IUPAC Name
propane-1,2,3-triol
Molecular Formula
C3H8O3
InChI Key
PEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILES
OCC(O)CO
Infrared spectrum
Conforms
Appearance (Form)
Clear viscous liquid
Color scale
=<10 APHA
GC
>=99.0 %
Refractive index
1.4700 to 1.4750 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text