Thermo Scientific Chemicals

2-Chloro-1,3,2-dioxaphospholane-2-oxide, 95%, Thermo Scientific Chemicals

Catalog number: 332061000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Chloro-1,3,2-dioxaphospholane-2-oxide, 95%, Thermo Scientific Chemicals

Catalog number: 332061000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 332061000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6609-64-9
IUPAC Name
2-chloro-1,3,2λ⁵-dioxaphospholan-2-one
Molecular Formula
C2H4ClO3P
InChI Key
SBMUNILHNJLMBF-UHFFFAOYSA-N
SMILES
ClP1(=O)OCCO1
Appearance (Form)
Liquid
Refractive index
1.4470 to 1.4530 (20°C, 589 nm)
Titration Argentometric
94 to 106 % (ex Cl)
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text