Thermo Scientific Chemicals

Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals

Catalog number: 332120025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals

Catalog number: 332120025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 332120025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-03-6
IUPAC Name
iodoethane
Molecular Formula
C2H5I
InChI Key
HVTICUPFWKNHNG-UHFFFAOYSA-N
SMILES
CCI
Stabilizer
silver
Appearance (Color)
Clear colorless to light yellow to pink
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text