Thermo Scientific Chemicals

Diethyl (2-aminoethyl)phosphonate oxalate, 97%, Thermo Scientific Chemicals

Catalog number: 332220010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Diethyl (2-aminoethyl)phosphonate oxalate, 97%, Thermo Scientific Chemicals

Catalog number: 332220010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 332220010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
203192-98-7
IUPAC Name
diethyl (2-azaniumylethyl)phosphonate
Molecular Formula
C6H17NO3P
InChI Key
JUAGHBASZWRMQH-UHFFFAOYSA-O
SMILES
CCOP(=O)(CC[NH3+])OCC
Titration with HClO4
>=96.0 %
Melting point
135°C to 141°C
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
Appearance (Color)
White to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text