Thermo Scientific Chemicals

Diethyl (4-nitrobenzyl)phosphonate, 98%, Thermo Scientific Chemicals

Catalog number: 332290250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Diethyl (4-nitrobenzyl)phosphonate, 98%, Thermo Scientific Chemicals

Catalog number: 332290250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 332290250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2609-49-6
IUPAC Name
diethyl [(4-nitrophenyl)methyl]phosphonate
Molecular Formula
C11H16NO5P
InChI Key
FORMFFDDQMCTCT-UHFFFAOYSA-N
SMILES
CCOP(=O)(CC1=CC=C(C=C1)[N+]([O-])=O)OCC
Infrared spectrum
Conforms
Refractive index
1.5220 to 1.5260 (20°C, 589 nm)
Appearance (Form)
Liquid
Appearance (Color)
Clear yellow to orange to dark red
NMR
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text