Thermo Scientific Chemicals

Carboxymethylecellulose, sodium salt, average M.W. 90000 (DS=0.7)

Catalog number: 332600010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Carboxymethylecellulose, sodium salt, average M.W. 90000 (DS=0.7)

Catalog number: 332600010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 332600010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9004-32-4
IUPAC Name
2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
Molecular Formula
(C12 H14 O9 R6)n
InChI Key
DPXJVFZANSGRMM-UHFFFAOYNA-N
SMILES
[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
Cadmium (Cd)
=<1 ppm
Degree of substitution
0.65 to 0.90
Appearance (Form)
Granular powder
Assay
>=99.5 %
Heavy metals
=<20 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text