Carboxymethylecellulose, sodium salt, average M.W. 250000 (DS=0.9)
Carboxymethylecellulose, sodium salt, average M.W. 250000 (DS=0.9)
Thermo Scientific Chemicals

Carboxymethylecellulose, sodium salt, average M.W. 250000 (DS=0.9)

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
View all
Glass bottle
Catalog number 332621000
Price (USD)/ Each
26.65
Online exclusive
29.40 
Save 2.75 (9%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
26.65
Online exclusive
29.40 
Save 2.75 (9%)
Add to cart
Carboxymethylecellulose, sodium salt, average M.W. 250000 (DS=0.9)
Catalog number332621000
Price (USD)/ Each
26.65
Online exclusive
29.40 
Save 2.75 (9%)
-
Add to cart
Chemical Identifiers
CAS9004-32-4
IUPAC Name2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
Molecular Formula(C12 H14 O9 R6)n
InChI KeyDPXJVFZANSGRMM-UHFFFAOYNA-N
SMILES[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Lead (Pb)=<3 ppm
Assay>=99.5 %
Heavy metals=<10 ppm
Arsenic (As)=<3 ppm
Mercury (Hg)=<1 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only