Carboxymethylecellulose, sodium salt, average M.W. 700000 (DS=0.9)
Carboxymethylecellulose, sodium salt, average M.W. 700000 (DS=0.9)
Thermo Scientific Chemicals

Carboxymethylecellulose, sodium salt, average M.W. 700000 (DS=0.9)

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
Packaging:
View all
Plastic bottle
Catalog number 332640010
Price (USD)/ Each
118.65
Online exclusive
132.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
118.65
Online exclusive
132.00 
Save 13.35 (10%)
Add to cart
Carboxymethylecellulose, sodium salt, average M.W. 700000 (DS=0.9)
Catalog number332640010
Price (USD)/ Each
118.65
Online exclusive
132.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9004-32-4
IUPAC Name2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
Molecular Formula(C12 H14 O9 R6)n
InChI KeyDPXJVFZANSGRMM-UHFFFAOYNA-N
SMILES[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=99.5 %
Arsenic (As)=<3 ppm
Lead (Pb)=<10 ppm
Cadmium (Cd)=<1 ppm
Impurity=<0.4 % (Sodium glycolate)
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only