Thermo Scientific Chemicals

Methyl 3-bromobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: 333650050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 3-bromobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: 333650050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 333650050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
618-89-3
IUPAC Name
methyl 3-bromobenzoate
Molecular Formula
C8H7BrO2
InChI Key
KMFJVYMFCAIRAN-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC(Br)=C1
Appearance (Form)
Low melting crystalline solid or liquid
HPLC
>=97.5 %
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text