Thermo Scientific Chemicals

4,6-Diaminoresorcinol dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 333670050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4,6-Diaminoresorcinol dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 333670050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 333670050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
16523-31-2
IUPAC Name
4,6-diaminobenzene-1,3-diol
Molecular Formula
C6H8N2O2
InChI Key
DPYROBMRMXHROQ-UHFFFAOYSA-N
SMILES
NC1=CC(N)=C(O)C=C1O
Appearance (Form)
Crystalline solid
Titration with NaOH
>=97.5 %
Appearance (Color)
Grey-brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text