Thermo Scientific Chemicals

Diethyl methylphosphonate, 96%, Thermo Scientific Chemicals

Catalog number: 333680050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Diethyl methylphosphonate, 96%, Thermo Scientific Chemicals

Catalog number: 333680050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 333680050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
683-08-9
IUPAC Name
diethyl methylphosphonate
Molecular Formula
C5H13O3P
InChI Key
NYYLZXREFNYPKB-UHFFFAOYSA-N
SMILES
CCOP(C)(=O)OCC
Infrared spectrum
Conforms
GC
>=95 %
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to light yellow
Refractive index
1.4130 to 1.4150 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text