Diethyl methylphosphonate, 96%, Thermo Scientific Chemicals
Diethyl methylphosphonate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl methylphosphonate, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 333680250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS683-08-9
IUPAC Namediethyl methylphosphonate
Molecular FormulaC5H13O3P
InChI KeyNYYLZXREFNYPKB-UHFFFAOYSA-N
SMILESCCOP(C)(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=95 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Refractive index1.4130 to 1.4150 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only