Chemical Identifiers
CAS58970-76-6
IUPAC Name(2R)-2-[(2S,3R)-3-amino-2-hydroxy-4-phenylbutanamido]-4-methylpentanoic acid
Molecular FormulaC16H24N2O4
InChI KeyVGGGPCQERPFHOB-MCIONIFRSA-N
SMILESCC(C)C[C@@H](NC(=O)[C@@H](O)[C@H](N)CC1=CC=CC=C1)C(O)=O
View more