6-Methoxy-1-indanone, 98%, Thermo Scientific Chemicals
6-Methoxy-1-indanone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Methoxy-1-indanone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
View all
Glass Bottle
Catalog number 334070250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS13623-25-1
IUPAC Name6-methoxy-2,3-dihydro-1H-inden-1-one
Molecular FormulaC10H10O2
InChI KeyUJGDLLGKMWVCPT-UHFFFAOYSA-N
SMILESCOC1=CC=C2CCC(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point107°C to 111°C
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Color)White to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only