Thermo Scientific Chemicals

2-Hydroxyethyl acrylate, 97%, stabilized, Thermo Scientific Chemicals

Catalog number: 334082500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

2-Hydroxyethyl acrylate, 97%, stabilized, Thermo Scientific Chemicals

Catalog number: 334082500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 334082500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
818-61-1
IUPAC Name
2-hydroxyethyl prop-2-enoate
Molecular Formula
C5H8O3
InChI Key
OMIGHNLMNHATMP-UHFFFAOYSA-N
SMILES
OCCOC(=O)C=C
Infrared spectrum
Conforms
Stabilizer
200 to 450 ppm MEHQ
Appearance (Form)
Clear liquid
Color scale
=<30 APHA
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text