Thermo Scientific Chemicals

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%, Thermo Scientific Chemicals

Catalog number: 334120010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%, Thermo Scientific Chemicals

Catalog number: 334120010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 334120010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
22572-40-3
IUPAC Name
(3-{[(ethylimino)methylidene]amino}propyl)trimethylazanium iodide
Molecular Formula
C9H20IN3
InChI Key
AGSKWMRPXWHSPF-UHFFFAOYSA-M
SMILES
[I-].CCN=C=NCCC[N+](C)(C)C
Infrared spectrum
Conforms
Titration Argentometric
>=97.5 %
Appearance (Form)
Crystalline powder
Appearance (Color)
White to off-white
Melting point
92°C to 97°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text