Thermo Scientific Chemicals

Polysorbate 85, Thermo Scientific Chemicals

Catalog number: 334152500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

Polysorbate 85, Thermo Scientific Chemicals

Catalog number: 334152500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 334152500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
9005-70-3
Molecular Formula
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC42H76O7
InChI Key
ZGEQUSWSYZZNAI-CLFAGFIQNA-N
SMILES
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)C1OCC(OCCO)C1OCCO
MDL Number
MFCD01779641
Appearance (Color)
Yellow to amber
Water
=<3 %
Hydroxyl value
38 to 52 mg KOH/g
Infrared spectrum
Conforms
Acid value
=<2 mg KOH/g

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text