Piperonyl butoxide, 90%, Tech., Thermo Scientific Chemicals
Piperonyl butoxide, 90%, Tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Piperonyl butoxide, 90%, Tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 334161000
Price (USD)/ Each
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name5-{[2-(2-butoxyethoxy)ethoxy]methyl}-6-propyl-2H-1,3-benzodioxole
Molecular FormulaC19H30O5
InChI KeyFIPWRIJSWJWJAI-UHFFFAOYSA-N
SMILESCCCCOCCOCCOCC1=CC2=C(OCO2)C=C1CCC
Molecular Weight (g/mol)338.44
View more
SpecificationsSpecification SheetSpecification Sheet
Specific gravity(20°C) 1.055 to 1.065
Appearance (Form)Liquid
Refractive index1.4970 to 1.5070 (20°C, 589 nm)
Appearance (Color)Clear yellow
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only