Thermo Scientific Chemicals

cis-2-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 334270010
1 g, Glass Bottle, Each
Thermo Scientific Chemicals

cis-2-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 334270010
1 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 334270010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
63216-49-9
IUPAC Name
(1R,2S)-2-{[(tert-butoxy)carbonyl]amino}cyclohexane-1-carboxylate
Molecular Formula
C12H20NO4
InChI Key
QJEQJDJFJWWURK-BDAKNGLRSA-M
SMILES
CC(C)(C)OC(=O)N[C@H]1CCCC[C@H]1C([O-])=O
Appearance (Color)
White
Infrared spectrum
Conforms
Melting point
127°C to 131°C
Titration with NaOH
>=97.5 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text