2-Naphthalenethiol, 98+%, Thermo Scientific Chemicals
2-Naphthalenethiol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Naphthalenethiol, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 334520250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namenaphthalene-2-thiol
Molecular FormulaC10H8S
InChI KeyRFCQDOVPMUSZMN-UHFFFAOYSA-N
SMILESSC1=CC=C2C=CC=CC2=C1
Molecular Weight (g/mol)160.23
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige to light grey
Infrared spectrumConforms
Appearance (Form)Powder or crystals or flakes
Melting point79°C to 83°C
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only