Thermo Scientific Chemicals

Methyl phenylpropiolate, 98%, Thermo Scientific Chemicals

Catalog number: 334590050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl phenylpropiolate, 98%, Thermo Scientific Chemicals

Catalog number: 334590050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 334590050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4891-38-7
IUPAC Name
methyl 3-phenylprop-2-ynoate
Molecular Formula
C10H8O2
InChI Key
JFGWPXKGINUNDH-UHFFFAOYSA-N
SMILES
COC(=O)C#CC1=CC=CC=C1
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.5615 to 1.5645 (20°C, 589 nm)
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text