Methyl phenylpropiolate, 98%, Thermo Scientific Chemicals
Methyl phenylpropiolate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl phenylpropiolate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 334590250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4891-38-7
IUPAC Namemethyl 3-phenylprop-2-ynoate
Molecular FormulaC10H8O2
InChI KeyJFGWPXKGINUNDH-UHFFFAOYSA-N
SMILESCOC(=O)C#CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5615 to 1.5645 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only