Sodium caprylate, 98%
Sodium caprylate, 98%
Sodium caprylate, 98%
Sodium caprylate, 98%
Sodium caprylate, 98%
Thermo Scientific Chemicals

Sodium caprylate, 98%

CAS: 1984-06-1 | C8H15NaO2 | 166.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 334600025
Price (USD)
-
Quantity:
2.5 kg
Request bulk or custom format
Chemical Identifiers
CAS1984-06-1
IUPAC Namesodium octanoate
Molecular FormulaC8H15NaO2
InChI KeyBYKRNSHANADUFY-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=97.5 % (on dry substance)
Appearance (Form)Powder
Appearance (Color)White to off-white
Water=<5 % (K.F.)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link