Natriumcaprylat, 98 %, Thermo Scientific Chemicals
Natriumcaprylat, 98 %, Thermo Scientific Chemicals
Natriumcaprylat, 98 %, Thermo Scientific Chemicals
Natriumcaprylat, 98 %, Thermo Scientific Chemicals
Natriumcaprylat, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Natriumcaprylat, 98 %, Thermo Scientific Chemicals

CAS: 1984-06-1 | C8H15NaO2 | 166.2 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
100 g
500 g
2.5 kg
Katalognummer 334601000
Preis (EUR)
44,88
Exklusiv online
52,80
Ersparnis 7,92 (15%)
Each
-
Zum Warenkorb hinzufügen
Menge:
100 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
44,88
Exklusiv online
52,80
Ersparnis 7,92 (15%)
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS1984-06-1
IUPAC Namesodium octanoate
Molecular FormulaC8H15NaO2
InChI KeyBYKRNSHANADUFY-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCC([O-])=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Infrared spectrumConforms
Titration with HClO4>=97.5 % (on dry substance)
Appearance (Form)Powder
Appearance (Color)White to off-white
Water=<5 % (K.F.)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Dokumente und Downloads

Zertifikate

Chargen-Nr.Certificate TypeDateCatalog Number(s)
A0471305Certificate of Analysis17. Mai 2025334600025, 334605000, 334601000
A0468437Certificate of Analysis31. Jan. 2025334600025, 334605000, 334601000
A0451912Certificate of Analysis02. Mai 2023334600025, 334605000, 334601000
A0393821Certificate of Analysis14. Dez. 2022334600025, 334605000, 334601000
A0410214Certificate of Analysis13. Dez. 2022334600025, 334605000, 334601000
5 Ergebnisse angezeigt, oben nach einem bestimmten Zertifikat suchen

Sicherheitsdatenblätter

Sicherheit und Handhabung

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Warning

Hazard Statements

H315 - Causes skin irritation

H319 - Causes serious eye irritation

H335 - May cause respiratory irritation

EU Specific Hazard Statements

May form combustible dust concentrations in air

Precautionary Statements

P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

Geben Sie die Katalognummer, den Namen oder Link frei.

1x1 image pixel for data collection