Thermo Scientific Chemicals

N-BOC-O-Benzyl-D-serine, 98%, Thermo Scientific Chemicals

Catalog number: 334670050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

N-BOC-O-Benzyl-D-serine, 98%, Thermo Scientific Chemicals

Catalog number: 334670050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 334670050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
47173-80-8
IUPAC Name
(2R)-3-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}propanoate
Molecular Formula
C15H20NO5
InChI Key
DMBKPDOAQVGTST-GFCCVEGCSA-M
SMILES
CC(C)(C)OC(=O)N[C@H](COCC1=CC=CC=C1)C([O-])=O
Appearance (Color)
White
Appearance (Form)
Powder
Melting point
58°C to 63°C
HPLC
>=97.5 %
Specific optical rotation
-20° to -24° (20°C, 589 nm) (c=1, EtOH)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text