N-BOC-O-Benzyl-D-serine, 98%, Thermo Scientific Chemicals
N-BOC-O-Benzyl-D-serine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-BOC-O-Benzyl-D-serine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 334670250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name(2R)-3-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}propanoate
Molecular FormulaC15H20NO5
InChI KeyDMBKPDOAQVGTST-GFCCVEGCSA-M
SMILESCC(C)(C)OC(=O)N[C@H](COCC1=CC=CC=C1)C([O-])=O
Molecular Weight (g/mol)294.33
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point58°C to 63°C
HPLC>=97.5 %
Specific optical rotation-20° to -24° (20°C, 589 nm) (c=1, EtOH)
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only