Thermo Scientific Chemicals

Docosanoic acid, 85%, technical, Thermo Scientific Chemicals

Catalog number: 334810010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Docosanoic acid, 85%, technical, Thermo Scientific Chemicals

Catalog number: 334810010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 334810010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-85-6
IUPAC Name
docosanoic acid
Molecular Formula
C22H44O2
InChI Key
UKMSUNONTOPOIO-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCCCCC(O)=O
GC
>=80 %
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or flakes or granules
Appearance (Color)
White to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text