Thermo Scientific Chemicals

3-Hydroxy-2,4,6-tribromobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: 334890250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Hydroxy-2,4,6-tribromobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: 334890250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 334890250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14348-40-4
IUPAC Name
2,4,6-tribromo-3-hydroxybenzoate
Molecular Formula
C7H2Br3O3
InChI Key
YDBHVMTTYXWHLI-UHFFFAOYSA-M
SMILES
OC1=C(Br)C=C(Br)C(C([O-])=O)=C1Br
Appearance (Form)
Powder or crystals
Melting point
144°C to 149°C
Infrared spectrum
Conforms
Appearance (Color)
White to light yellow
Titration with NaOH
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text