1,2-Benzenedithiol, 96%, Thermo Scientific Chemicals
1,2-Benzenedithiol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2-Benzenedithiol, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 334920050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS17534-15-5
IUPAC Namebenzene-1,2-dithiol
Molecular FormulaC6H6S2
InChI KeyJRNVQLOKVMWBFR-UHFFFAOYSA-N
SMILESSC1=C(S)C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=95 %
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid or solid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only