2-Amino-4,5-dimethoxybenzoic acid, 98%, Thermo Scientific Chemicals
2-Amino-4,5-dimethoxybenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-4,5-dimethoxybenzoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 335010250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5653-40-7
IUPAC Name2-amino-4,5-dimethoxybenzoic acid
Molecular FormulaC9H11NO4
InChI KeyHJVAVGOPTDJYOJ-UHFFFAOYSA-N
SMILESCOC1=CC(N)=C(C=C1OC)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to grey to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=97.5 % (On dry substance)
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only