Thermo Scientific Chemicals

2-Amino-4,5-dimethoxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 335011000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Amino-4,5-dimethoxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 335011000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 335011000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5653-40-7
IUPAC Name
2-amino-4,5-dimethoxybenzoic acid
Molecular Formula
C9H11NO4
InChI Key
HJVAVGOPTDJYOJ-UHFFFAOYSA-N
SMILES
COC1=CC(N)=C(C=C1OC)C(O)=O
Appearance (Color)
White to beige to grey to brown
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with NaOH
>=97.5 % (On dry substance)
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text