Thermo Scientific Chemicals

1-Allylimidazole, 97%, Thermo Scientific Chemicals

Catalog number: 335170250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Allylimidazole, 97%, Thermo Scientific Chemicals

Catalog number: 335170250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 335170250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
31410-01-2
IUPAC Name
1-(prop-2-en-1-yl)-1H-imidazole
Molecular Formula
C6H8N2
InChI Key
XLXCHZCQTCBUOX-UHFFFAOYSA-N
SMILES
C=CCN1C=CN=C1
Appearance (Color)
Clear colorless to yellow to light orange
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=96.0 %
Refractive index
1.5030 to 1.5060 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text