Thermo Scientific Chemicals

2,4,6-Trichlorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 335180250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2,4,6-Trichlorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 335180250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 335180250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4136-95-2
IUPAC Name
2,4,6-trichlorobenzoyl chloride
Molecular Formula
C7H2Cl4O
InChI Key
OZGSEIVTQLXWRO-UHFFFAOYSA-N
SMILES
ClC(=O)C1=C(Cl)C=C(Cl)C=C1Cl
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=97.5 %
Refractive index
1.5750 to 1.5770 (20°C, 589 nm)
Appearance (Color)
Clear yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text