Thermo Scientific Chemicals

Lithium azide, pure, 20% solution in water, Thermo Scientific Chemicals

Catalog number: 335361000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Lithium azide, pure, 20% solution in water, Thermo Scientific Chemicals

Catalog number: 335361000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 335361000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19597-69-4
IUPAC Name
lithium(1+) azide
Molecular Formula
LiN3
InChI Key
GUWHRJQTTVADPB-UHFFFAOYSA-N
SMILES
[Li+].[N-]=[N+]=[N-]
Appearance (Form)
Solution
Appearance (Color)
Clear colorless
Titration Argentometric
17 to 23 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, For Azidation/Diazo transfer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text