Zirconium(IV) ethoxide, 99%, Thermo Scientific Chemicals
Zirconium(IV) ethoxide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium(IV) ethoxide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 335450050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS18267-08-8
IUPAC Namezirconium(4+) ethanolate
Molecular FormulaC2H5OZr
InChI KeyVCYRFVPNTBNCLW-UHFFFAOYSA-N
SMILES[Zr+4].CC[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder and/or chunks
Infrared spectrumConforms
Assay>=98.5 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only