Thermo Scientific Chemicals

2-(Dimethylaminomethyl)-1-cyclohexanone hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 335620010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

2-(Dimethylaminomethyl)-1-cyclohexanone hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 335620010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 335620010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
42036-65-7
IUPAC Name
hydrogen 2-[(dimethylamino)methyl]cyclohexan-1-one chloride
Molecular Formula
C9H18ClNO
InChI Key
CLVHTSWMNNSUSH-UHFFFAOYNA-N
SMILES
[H+].[Cl-].CN(C)CC1CCCCC1=O
Titration with HClO4
>=97.5 %
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text