Thermo Scientific Chemicals

(+)-Diacetyl-L-tartaric anhydride, 97%, Thermo Scientific Chemicals

Catalog number: 336040250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

(+)-Diacetyl-L-tartaric anhydride, 97%, Thermo Scientific Chemicals

Catalog number: 336040250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 336040250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6283-74-5
IUPAC Name
(3R,4R)-4-(acetyloxy)-2,5-dioxooxolan-3-yl acetate
Molecular Formula
C8H8O7
InChI Key
XAKITKDHDMPGPW-IOMOGOHMNA-N
SMILES
CC(=O)O[C@@H]1[C@@H](OC(C)=O)C(=O)OC1=O
Appearance (Color)
White
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals or needles
Melting point
130°C to 135°C
Titration with NaOH
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text