Diethyl allylphosphonate, 97%, Thermo Scientific Chemicals
Diethyl allylphosphonate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl allylphosphonate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 336110250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1067-87-4
IUPAC Namediethyl (prop-2-en-1-yl)phosphonate
Molecular FormulaC7H15O3P
InChI KeyYPJHXRAHMUKXAE-UHFFFAOYSA-N
SMILESCCOP(=O)(CC=C)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4325 to 1.4345 (20°C, 589 nm)
Appearance (Color)Clear colorless
GC>=96.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only